Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133204.2 | NP_573467.2 | 6 | 1317 | ||
GenBank mRNA | AF204776.1 | - | 1 | 191 | ||
AK139659.1 | - | 6 | 1318 | |||
BC104342.1 | - | 1 | 147 | |||
BC104343.1 | - | 1 | 147 | |||
BC139442.1 | - | 5 | 1207 | 8862464 | ||
BC139456.1 | - | 5 | 1207 | 8862478 |