Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168621.1 | NP_001162092.1 | 5 | 1410 | ||
NM_001168622.1 | NP_001162093.1 | 3 | 2425 | |||
NM_001168623.1 | NP_001162094.1 | 5 | 609 | |||
NM_133206.3 | NP_573469.1 | 4 | 2521 | |||
GenBank mRNA | AF378525.1 | - | 5 | 1115 | ||
AK030575.1 | - | 3 | 1571 | |||
AK076756.1 | - | 4 | 1668 | |||
AK078819.1 | - | 5 | 636 | |||
AK079338.1 | - | 5 | 1410 | |||
AK191998.1 | - | 1 | 218 | |||
BC006765.1 | - | 4 | 2121 | 3710377 | ||
BC086770.1 | - | 4 | 2319 | 5720382 |