Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pilrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133209.2 | NP_573472.2 | 2 | 534 | ||
GenBank mRNA | AB122024.2 | - | 2 | 429 | ||
AJ400847.1 | - | 2 | 483 | |||
BC145880.1 | - | 2 | 472 | 40131183 | ||
BC145906.1 | - | 2 | 472 | 40131209 | ||
BC172053.1 | - | 2 | 472 | 9054339 |