Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133240.2 | NP_573503.2 | 1, 2 | 202 | ||
GenBank mRNA | AF441166.1 | - | 1, 2 | 202 | ||
AK033724.1 | - | 1, 2 | 171 | |||
AK151924.1 | - | 1, 2 | 180 | |||
BC005792.1 | - | 1, 2 | 202 | 3603527 |