Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 3 | 314 | ||
NM_001252486.1 | NP_001239415.1 | 3 | 314 | |||
NM_133247.3 | NP_573510.2 | 3 | 314 | |||
GenBank mRNA | AF383174.1 | - | 3 | 303 | ||
AK047530.1 | - | 3 | 281 | |||
AK053227.1 | - | 3 | 265 | |||
AK082251.1 | - | 3 | 297 | |||
AK082943.1 | - | 3 | 297 | |||
AK087467.1 | - | 3 | 315 | |||
AK122442.1 | - | 2 | 145 | |||
AK143844.1 | - | 3 | 282 | |||
BC005506.1 | - | 3 | 267 | |||
BC089315.1 | - | 3 | 260 | |||
BY731439.1 | - | 3 | 311 |