Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133187.3 | NP_573450.2 | 1 | 593 | ||
GenBank mRNA | AF285091.1 | - | 1 | 535 | ||
AK053058.1 | - | 1 | 467 | |||
AK143522.1 | - | 1 | 324 | |||
BC056975.1 | - | 1 | 362 | 6827227 | ||
EU797522.1 | - | 1 | 371 |