Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133216.3 | NP_573479.3 | 7 | 687 | ||
GenBank mRNA | AF363970.1 | - | 7 | 627 | ||
AK146033.1 | - | 7 | 667 | |||
AK149676.1 | - | 7 | 626 | |||
AK158597.1 | - | 5 | 645 | |||
AK166715.1 | - | 7 | 587 | |||
AK169024.1 | - | 6 | 513 | |||
AK181539.1 | - | 2 | 96 | |||
AK181745.1 | - | 96 | ||||
BC065174.1 | - | 7 | 575 | 6415314 |