Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133226.2 | NP_573489.2 | 4 | 185 | ||
GenBank mRNA | AF334612.1 | - | 4 | 181 | ||
AK052748.1 | - | 4 | 186 |