Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133241.2 | NP_573504.1 | 12 | 1145 | ||
GenBank mRNA | AF449425.1 | - | 12 | 1599 | ||
AK030396.1 | - | 12 | 1144 | |||
AK043757.1 | - | 12 | 1145 | |||
AK122190.1 | - | 12 | 1587 | |||
BC024719.1 | - | 12 | 1145 | 5363238 |