Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 4, 5 | 419 | ||
NM_001252486.1 | NP_001239415.1 | 4, 5 | 419 | |||
NM_133247.3 | NP_573510.2 | 4, 5 | 419 | |||
GenBank mRNA | AF383174.1 | - | 4, 5 | 408 | ||
AK047530.1 | - | 4, 5 | 381 | |||
AK053227.1 | - | 4, 5 | 370 | |||
AK082251.1 | - | 4, 5 | 402 | |||
AK082943.1 | - | 4, 5 | 402 | |||
AK087467.1 | - | 4, 5 | 420 | |||
AK122442.1 | - | 3, 4 | 250 | |||
AK143844.1 | - | 4, 5 | 387 | |||
BC005506.1 | - | 4, 5 | 372 | |||
BC067399.1 | - | 3, 4 | 261 | |||
BC089315.1 | - | 4, 5 | 365 | |||
BY731439.1 | - | 4, 5 | 416 |