Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277219.1 | NP_001264148.1 | 2 | 538 | ||
NM_133656.5 | NP_598417.2 | 2 | 538 | |||
GenBank mRNA | AF239673.1 | - | 2 | 408 | ||
AK028179.1 | - | 2 | 515 | |||
AK028488.1 | - | 2 | 488 | |||
AK036905.1 | - | 2 | 489 | |||
AK052795.1 | - | 2 | 497 | |||
AK076097.1 | - | 2 | 233 | |||
AK088519.1 | - | 2 | 509 | |||
AK139089.1 | - | 2 | 494 | |||
AK152549.1 | - | 2 | 512 | |||
AK154987.1 | - | 2 | 523 | |||
AK161356.1 | - | 2 | 488 | |||
AK161761.1 | - | 2 | 480 | |||
BC012216.1 | - | 2 | 474 | 3992341 | ||
BC031149.1 | - | 2 | 471 | 4975654 | ||
S72408.1 | - | 2 | 463 |