Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133657.1 | NP_598418.1 | 2 | 333 | ||
GenBank mRNA | BC018356.1 | - | 2 | 333 | 4195640 | |
BC094017.1 | - | 2 | 321 | 5101743 | ||
L06463.1 | - | 2 | 319 |
Gene Symbol: Cyp2a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101467.1 | NP_001094937.1 | 2 | 344 | ||
GenBank mRNA | AK149509.1 | - | 2 | 343 | ||
BC151116.1 | - | 2 | 338 | 9088029 | ||
BC158034.1 | - | 2 | 338 | 9007337 |