Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130903.3 | NP_570973.1 | 1544 | |||
GenBank mRNA | AF373410.1 | - | 1544 |