Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085409.1 | NP_001078878.1 | 6 | 1698 | ||
NM_133186.3 | NP_573449.2 | 5 | 1325 | |||
GenBank mRNA | AK028961.1 | - | 5 | 1302 | ||
AK078769.1 | - | 5 | 1719 | |||
AK167028.1 | - | 6 | 1698 | |||
AK171237.1 | - | 5 | 1330 | |||
AK217925.1 | - | 3 | 407 | |||
AY029586.1 | - | 5 | 1313 | |||
AY214462.1 | - | 1691 | ||||
BC027251.1 | - | 5 | 1308 | |||
BC037435.1 | - | 5 | 1326 | 2651584 |