Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289730.1 | NP_001276659.1 | 16, 17 | 1971 | ||
NM_001289731.1 | NP_001276660.1 | 18, 19 | 2479 | |||
NM_133197.4 | NP_573460.2 | 15, 16 | 1923 | |||
GenBank mRNA | AB052945.1 | - | 15, 16 | 1866 | ||
AB101616.1 | - | 16, 17 | 1914 | |||
AB244989.1 | - | 6, 7 | 477 | |||
AK032122.1 | - | 15, 16 | 1924 | |||
AK045424.1 | - | 18, 19 | 2481 | |||
BC125435.1 | - | 15, 16 | 1825 | 40129950 | ||
X57298.1 | - | 4, 5 | 304 |