Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133239.2 | NP_573502.2 | 9 | 3481 | ||
GenBank mRNA | AF406641.1 | - | 9 | 3446 | ||
AF492496.1 | - | 8 | 3322 | |||
AK164413.1 | - | 1 | 445 | |||
BC031378.1 | - | 2 | 541 | |||
BC053331.1 | - | 4 | 1724 |