Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 5, 6 | 572 | ||
NM_001252486.1 | NP_001239415.1 | 5, 6 | 572 | |||
NM_133247.3 | NP_573510.2 | 5, 6 | 572 | |||
GenBank mRNA | AF383174.1 | - | 5, 6 | 561 | ||
AK047530.1 | - | 5, 6 | 534 | |||
AK053227.1 | - | 5, 6 | 523 | |||
AK082251.1 | - | 5, 6 | 555 | |||
AK082943.1 | - | 5, 6 | 555 | |||
AK087467.1 | - | 5, 6 | 573 | |||
AK122442.1 | - | 4, 5 | 403 | |||
AK143844.1 | - | 5, 6 | 540 | |||
BC005506.1 | - | 5, 6 | 525 | |||
BC067399.1 | - | 4, 5 | 414 | |||
BC089315.1 | - | 5, 6 | 518 |