Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133354.2 | NP_579932.1 | 4 | 357 | ||
GenBank mRNA | AK012619.1 | - | 4 | 358 | ||
AK085238.1 | - | 4 | 304 | |||
AK132213.1 | - | 356 | ||||
AK160404.1 | - | 4 | 374 | |||
AK160418.1 | - | 4 | 376 | |||
AK167308.1 | - | 4 | 367 | |||
AK219519.1 | - | 2 | 65 | |||
BC017522.1 | - | 4 | 331 | 4512364 | ||
BC083326.1 | - | 4 | 269 | 6819700 | ||
BC095930.1 | - | 4 | 344 | 4936947 | ||
L79948.1 | - | 4 | 342 |