Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133485.2 | NP_597844.1 | 4 | 2224 | ||
GenBank mRNA | AF407167.2 | - | 4 | 2320 | ||
AK085997.1 | - | 4 | 2262 | |||
BC055717.1 | - | 4 | 1831 | 6415732 |