Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222954.1 | - | 4 | 555 | ||
AK223210.1 | - | 4 | 560 | |||
AK313583.1 | - | 4 | 552 | |||
AX775815.1 | - | 4 | 551 | |||
AX775817.1 | - | 4 | 548 | |||
BC002975.1 | - | 4 | 535 | 3542354 | ||
BC032651.1 | - | 4 | 518 | 5504755 | ||
M80461.1 | - | 4 | 498 | |||
M89957.1 | - | 4 | 576 | |||
S52229.1 | - | 4 | 482 | |||
S79249.1 | - | 3 | 170 | |||
X83539.1 | - | 3 | 170 |