Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111059.1 | NP_001104529.1 | 5 | 762 | ||
NM_133654.3 | NP_598415.1 | 5 | 762 | |||
GenBank mRNA | AK044121.1 | - | 5 | 698 | ||
AK053874.1 | - | 5 | 698 | |||
AK089010.1 | - | 5 | 668 | |||
AK167329.1 | - | 5 | 673 | |||
AK167350.1 | - | 5 | 673 | |||
BC006607.1 | - | 5 | 678 | 3966337 | ||
BC066820.1 | - | 5 | 635 | |||
BU058977.1 | - | 3 | 245 | |||
S69293.1 | - | 5 | 640 | |||
S69299.1 | - | 5 | 617 | |||
S69301.1 | - | 5 | 617 |