Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133665.3 | NP_598426.1 | 4 | 685 | ||
GenBank mRNA | AK141270.1 | - | 4 | 665 | ||
AK155778.1 | - | 4 | 686 | |||
BC011070.1 | - | 4 | 653 | 3497896 | ||
S68893.1 | - | 4 | 507 |