Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083901.1 | NP_001077370.1 | 4, 5 | 541 | ||
NM_133680.3 | NP_598441.3 | 3, 4 | 638 | |||
GenBank mRNA | AK146596.1 | - | 4, 5 | 534 | ||
BC033469.1 | - | 3, 4 | 600 | 4951631 |