Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133683.3 | NP_598444.1 | 4 | 583 | ||
GenBank mRNA | AK034298.1 | - | 4 | 589 | ||
AK036228.1 | - | 3 | 660 | |||
AK042426.1 | - | 3 | 638 | |||
AK079735.1 | - | 441 | ||||
AK136778.1 | - | 572 | ||||
AK138931.1 | - | 4 | 571 | |||
AK155275.1 | - | 4 | 544 | |||
AK170143.1 | - | 4 | 543 | |||
AK171087.1 | - | 4 | 543 | |||
AK193317.1 | - | 1 | 47 | |||
BC016895.1 | - | 4 | 585 | 4239007 |