Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133716.3 | NP_598477.2 | 10 | 2250 | ||
GenBank mRNA | AB029329.1 | - | 1 | 658 | ||
AK007666.1 | - | 2 | 851 | |||
AK179675.1 | - | 1 | 246 | |||
BC031581.1 | - | 10 | 2239 | |||
BC032217.1 | - | 11 | 2095 | |||
BC051481.1 | - | 1 | 152 | |||
BC052413.1 | - | 10 | 2248 | 5720177 |