Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 2, 3 | 573 | ||
GenBank mRNA | AK008408.1 | - | 2, 3 | 459 | ||
AK077065.1 | - | 2, 3 | 576 | |||
AK150071.1 | - | 2, 3 | 476 | |||
AK159364.1 | - | 2, 3 | 500 | |||
AK161096.1 | - | 2, 3 | 455 | |||
AK170171.1 | - | 2, 3 | 476 | |||
AK170697.1 | - | 2, 3 | 477 | |||
AK178013.1 | - | 2, 3 | 213 | |||
AK192822.1 | - | 2, 3 | 213 | |||
AK198710.1 | - | 2, 3 | 214 | |||
AK200429.1 | - | 2, 3 | 237 | |||
AK203327.1 | - | 2, 3 | 214 | |||
BC018367.1 | - | 2, 3 | 425 | 2648510 | ||
BC018491.1 | - | 2, 3 | 453 | 2651602 | ||
BC026136.1 | - | 1, 2 | 49 |