Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133722.2 | NP_598483.2 | 2 | 770 | ||
GenBank mRNA | AK211213.1 | - | 1 | 18 | ||
BC018511.1 | - | 2 | 770 | 3483379 | ||
BM943880.1 | - | 2 | 251 |