Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133728.3 | NP_598489.2 | 4 | 560 | ||
GenBank mRNA | AK008875.1 | - | 4 | 477 | ||
AK031295.1 | - | 4 | 518 | |||
AK034746.1 | - | 479 | ||||
AK049194.1 | - | 4 | 507 | |||
AK050045.1 | - | 4 | 499 | |||
AK076793.1 | - | 4 | 462 | |||
AK154984.1 | - | 5 | 657 | |||
AK155030.1 | - | 4 | 499 | |||
AK170510.1 | - | 4 | 476 | |||
AK206632.1 | - | 1 | 28 | |||
BC013617.1 | - | 4 | 380 | 3979452 |