Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 3 | 433 | ||
GenBank mRNA | AK150365.1 | - | 4 | 461 | ||
AK151044.1 | - | 3 | 401 | |||
AK152180.1 | - | 3 | 381 | |||
AK152423.1 | - | 3 | 346 | |||
AK153122.1 | - | 3 | 346 | |||
AK153378.1 | - | 4 | 461 | |||
AK153539.1 | - | 3 | 381 | |||
AK155767.1 | - | 3 | 401 | |||
AK157001.1 | - | 4 | 358 | |||
AK162610.1 | - | 3 | 429 | |||
AK165671.1 | - | 3 | 400 | |||
AK166444.1 | - | 3 | 346 | |||
AK170823.1 | - | 3 | 392 | |||
AK172701.1 | - | 3 | 362 | |||
AK201658.1 | - | 3 | 224 | |||
AK211851.1 | - | 1 | 88 | |||
BC010292.1 | - | 3 | 382 | 3153174 | ||
BC086319.1 | - | 3 | 390 | 5293409 |