Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271531.1 | NP_001258460.1 | 3 | 509 | ||
NM_001271532.1 | NP_001258461.1 | 2 | 376 | |||
NM_133748.2 | NP_598509.1 | 3 | 752 | |||
NM_178082.3 | NP_835183.1 | 3 | 516 | |||
GenBank mRNA | AF527631.1 | - | 3 | 486 | ||
AK040703.1 | - | 3 | 507 | |||
AK152076.1 | - | 3 | 482 | |||
AK154543.1 | - | 3 | 515 | |||
AK161256.1 | - | 3 | 520 | |||
AK165634.1 | - | 3 | 521 | |||
AK188429.1 | - | 59 | ||||
BC015288.1 | - | 3 | 466 | 4240711 | ||
BC023067.1 | - | 3 | 490 | 5357599 | ||
BC023874.1 | - | 2 | 408 | 5355039 | ||
BC024411.1 | - | 3 | 504 | 4971297 |