Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133765.4 | NP_598526.3 | 6 | 818 | ||
GenBank mRNA | AB041586.1 | - | 6 | 667 | ||
AK009859.1 | - | 3 | 259 | |||
AK140412.1 | - | 6 | 819 | |||
AK163641.1 | - | 6 | 787 | |||
BC026929.1 | - | 6 | 803 | 4194857 |