Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133766.3 | NP_598527.2 | 3 | 386 | ||
GenBank mRNA | AB158474.1 | - | 3 | 390 | ||
AK030074.1 | - | 3 | 387 | |||
AK032461.1 | - | 3 | 359 | |||
AK044000.1 | - | 3 | 380 | |||
AK044475.1 | - | 3 | 382 | |||
AK052176.1 | - | 3 | 366 | |||
AK052906.1 | - | 3 | 382 | |||
AK129065.1 | - | 3 | 364 | |||
AK161250.1 | - | 3 | 371 | |||
BC007482.1 | - | 3 | 390 | 3495608 | ||
BC032092.1 | - | 3 | 391 |