Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282118.1 | NP_001269047.1 | 2, 3 | 403 | ||
NM_001282119.1 | NP_001269048.1 | 2, 3 | 325 | |||
NM_001282120.1 | NP_001269049.1 | 2, 3 | 681 | |||
NM_133767.3 | NP_598528.2 | 2, 3 | 681 | |||
GenBank mRNA | AK010766.1 | - | 2, 3 | 626 | ||
AK136344.1 | - | 2, 3 | 365 | |||
AK146961.1 | - | 2, 3 | 621 | |||
BC016590.1 | - | 2, 3 | 257 | 3585359 | ||
BC066819.1 | - | 2, 3 | 575 | |||
CK620846.1 | - | 2, 3 | 314 |