Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252459.1 | NP_001239388.1 | 2, 3 | 517 | ||
NM_001252460.1 | NP_001239389.1 | 2, 3 | 572 | |||
NM_133769.3 | NP_598530.2 | 2, 3 | 280 | |||
GenBank mRNA | AB093288.2 | - | 1, 2 | 152 | ||
AF162472.1 | - | 1, 2 | 140 | |||
AF334144.1 | - | 2, 3 | 215 | |||
AK018280.1 | - | 2, 3 | 210 | |||
AK030397.1 | - | 2, 3 | 217 | |||
AK147224.1 | - | 2, 3 | 518 | |||
AK147586.1 | - | 2, 3 | 273 | |||
AK147632.1 | - | 2, 3 | 214 | |||
AK147971.1 | - | 2, 3 | 217 | |||
BC056974.1 | - | 2, 3 | 228 | 6831770 | ||
CJ115523.1 | - | 2, 3 | 280 |