Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000477.5 | NP_000468.1 | 6 | 723 | ||
GenBank mRNA | A06977.1 | - | 6 | 725 | ||
AF190168.1 | - | 6 | 650 | |||
AF542069.1 | - | 6 | 683 | |||
AK292755.1 | - | 6 | 688 | |||
AK298437.1 | - | 6 | 716 | |||
AK298461.1 | - | 6 | 433 | |||
AK308044.1 | - | 6 | 565 | |||
AK314794.1 | - | 6 | 691 | |||
AY550967.1 | - | 6 | 657 | |||
AY728024.1 | - | 6 | 656 | |||
AY960291.1 | - | 6 | 682 | |||
BC034023.1 | - | 6 | 691 | 4734617 | ||
BC036003.1 | - | 6 | 691 | 4724105 | ||
CR749331.1 | - | 6 | 690 | |||
DQ986150.1 | - | 6 | 661 | |||
HQ537426.1 | - | 5 | 578 | |||
V00494.1 | - | 6 | 689 | |||
V00495.1 | - | 6 | 725 |