Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133681.4 | NP_598442.1 | 6 | 713 | ||
GenBank mRNA | AK007428.1 | - | 6 | 711 | ||
AK078869.1 | - | 6 | 714 | |||
AK162521.1 | - | 6 | 713 | |||
BC003448.1 | - | 6 | 653 | 2654254 |