Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133683.3 | NP_598444.1 | 4, 5 | 618 | ||
GenBank mRNA | AK018383.1 | - | 4, 5 | 566 | ||
AK034298.1 | - | 4, 5 | 624 | |||
AK036228.1 | - | 3, 4 | 695 | |||
AK042426.1 | - | 3, 4 | 673 | |||
AK079735.1 | - | 476 | ||||
AK136778.1 | - | 607 | ||||
AK138931.1 | - | 4, 5 | 606 | |||
AK155275.1 | - | 4, 5 | 579 | |||
AK170143.1 | - | 4, 5 | 578 | |||
AK171087.1 | - | 4, 5 | 578 | |||
AK193317.1 | - | 1, 2 | 82 | |||
BC016895.1 | - | 4, 5 | 620 | 4239007 |