Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133692.2 | NP_598453.1 | 12 | 1809 | ||
GenBank mRNA | AK031778.1 | - | 12 | 1811 | ||
AK088385.1 | - | 12 | 1802 | |||
AK162240.1 | - | 7 | 1138 | |||
AK168179.1 | - | 12 | 1797 | |||
BC055325.1 | - | 12 | 1812 | 6402059 |