Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133693.2 | NP_598454.2 | 8 | 1204 | ||
GenBank mRNA | AK005626.1 | - | 5 | 644 | ||
AK013929.1 | - | 8 | 1145 | |||
BC009148.1 | - | 8 | 1141 | 3600406 | ||
BC011286.1 | - | 8 | 1123 | 3485410 | ||
BC027372.1 | - | 8 | 1128 | 4952304 | ||
BC039917.1 | - | 8 | 1139 | |||
BC057928.1 | - | 8 | 1130 | 5289543 |