Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133701.2 | NP_598462.1 | 3 | 447 | ||
GenBank mRNA | AK011639.1 | - | 3 | 389 | ||
AK081998.1 | - | 3 | 382 | |||
AK145461.1 | - | 3 | 450 | |||
AK166890.1 | - | 3 | 399 | |||
BC014869.1 | - | 3 | 374 | 3602158 | ||
BC023691.1 | - | 3 | 376 | 5344332 | ||
BC025030.1 | - | 3 | 393 | 4947416 | ||
BY735790.1 | - | 3 | 447 |