Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133710.3 | NP_598471.3 | 8 | 1119 | ||
GenBank mRNA | AJ344340.1 | - | 8 | 1201 | ||
AK013115.1 | - | 8 | 1115 | |||
AK142857.1 | - | 3 | 902 | |||
BC094289.1 | - | 8 | 804 | 4986338 |