Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 4 | 693 | ||
GenBank mRNA | AK008408.1 | - | 4 | 579 | ||
AK077065.1 | - | 4 | 696 | |||
AK150071.1 | - | 4 | 596 | |||
AK159364.1 | - | 4 | 620 | |||
AK161096.1 | - | 4 | 575 | |||
AK170171.1 | - | 4 | 596 | |||
AK170697.1 | - | 4 | 597 | |||
BC018367.1 | - | 4 | 545 | 2648510 | ||
BC018491.1 | - | 4 | 573 | 2651602 | ||
BC026136.1 | - | 3 | 169 |