Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163833.1 | NP_001157305.1 | 2 | 874 | ||
GenBank mRNA | AK006842.1 | - | 1 | 274 | ||
AK044179.1 | - | 1 | 275 | |||
AK155382.1 | - | 2 | 860 | |||
BC013191.1 | - | 1 | 476 | |||
BC037487.1 | - | 2 | 885 |