Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133762.3 | NP_598523.3 | 14 | 1670 | ||
GenBank mRNA | AY455829.1 | - | 8 | 781 | ||
BC008539.1 | - | 3 | 351 | |||
BC076631.1 | - | 13 | 1480 | 30637859 |