Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252459.1 | NP_001239388.1 | 3 | 541 | ||
NM_001252460.1 | NP_001239389.1 | 3 | 596 | |||
NM_133769.3 | NP_598530.2 | 3 | 304 | |||
GenBank mRNA | AB093288.2 | - | 2 | 176 | ||
AF162472.1 | - | 2 | 164 | |||
AF334144.1 | - | 3 | 239 | |||
AK018280.1 | - | 3 | 234 | |||
AK030397.1 | - | 3 | 241 | |||
AK147224.1 | - | 3 | 542 | |||
AK147586.1 | - | 3 | 297 | |||
AK147632.1 | - | 3 | 238 | |||
AK147971.1 | - | 3 | 241 | |||
BC056974.1 | - | 3 | 252 | 6831770 | ||
CJ115523.1 | - | 3 | 304 |