Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133667.2 | NP_598428.2 | 11 | 1964 | ||
GenBank mRNA | AF267660.1 | - | 11 | 1927 | ||
AK002647.1 | - | 11 | 1907 | |||
BC021764.1 | - | 11 | 1918 | 3488813 | ||
CX204954.1 | - | 1 | 468 |