Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000491.3 | NP_000482.3 | 3 | 471 | ||
XM_011542059.2 | XP_011540361.1 | 4 | 622 | |||
GenBank mRNA | BC008983.1 | - | 3 | 413 | 4212848 | |
BQ711602.1 | - | 4 | 633 | |||
X03084.1 | - | 2 | 265 |