Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026994.1 | NP_081270.1 | 8, 9 | 887 | ||
NM_133679.2 | NP_598440.1 | 9, 10 | 932 | |||
GenBank mRNA | AK008846.1 | - | 9, 10 | 705 | ||
AK145827.1 | - | 9, 10 | 756 | |||
AK170997.1 | - | 1, 2 | 48 | |||
BC010479.1 | - | 9, 10 | 729 | 3968727 | ||
BC019387.1 | - | 8, 9 | 696 | 4164847 |