Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083901.1 | NP_001077370.1 | 13 | 2564 | ||
NM_133680.3 | NP_598441.3 | 12 | 2661 | |||
GenBank mRNA | AK003654.1 | - | 1 | 584 | ||
AK027994.1 | - | 1 | 992 | |||
AK077464.1 | - | 1 | 759 | |||
AK146596.1 | - | 13 | 2557 | |||
BC018395.1 | - | 5 | 1450 | |||
BC033469.1 | - | 12 | 2624 | 4951631 |