Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000493.3 | NP_000484.2 | 2, 3 | 258 | ||
XM_006715333.3 | XP_006715396.1 | 2, 3 | 977 | |||
XM_011535432.2 | XP_011533734.1 | 2, 3 | 1905 | |||
XM_011535433.2 | XP_011533735.1 | 3, 4 | 2052 | |||
XM_017010248.1 | XP_016865737.1 | 1, 2 | 3705 | |||
XM_017010249.1 | XP_016865738.1 | 2, 3 | 5383 | |||
GenBank mRNA | BC130621.1 | - | 1, 2 | 188 | 40146652 | |
BC130623.1 | - | 1, 2 | 188 | 40146654 | ||
BI521533.1 | - | 2, 3 | 248 |